Order Entry
ContactUsLinkComponent
VWR* Traceable* Conductivity Calibration Standard
VWR® Traceable® Conductivity Calibration Standards (CRM)
Catalog # 62344-936
Supplier:  VWR International
CAS Number:  
undefined
VWR® Traceable® Conductivity Calibration Standards (CRM)
Catalog # 62344-936
Supplier:  VWR International
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    473 ml (16 oz.) bottles (NIST/ISO 34, ISO 17025/A2LA certificate)
  • Resistivity:
    0.00071 MΩ
  • Conductivity:
    1413 µmhos (µS)
  • Dissolved Solids:
    933 ppm

Specifications

About this item

These Certified Reference Materials (CRM) calibrate conductivity meters and probes for maximum accuracy.

  • Traceable
  • Available in single-use (100 ml) or 473 ml (16 oz.) sizes
  • Easy to calibrate
  • Polyethylene container
  • Ideal for both laboratory and field conditions

They are compatible with all makes of equipment, and can be used in laboratory or field.

Single-use standards make it easy to calibrate with no external contamination. An extra large opening of 4.4 cm (1³/₄") diameter and depth of 8.9 cm (3¹/₂") allow probe calibration to take place in the standard’s polyethylene container. Ideal for lab/field conditions.

All solutions include a serial numbered Traceable certificate to indicate traceability to standards provided by the National Institute of Standards and Technology (NIST). Double A2LA accreditation accredited certification provides the highest achievable level of quality assurance, documentation, and accuracy. CRMs are produced by A2LA accredited ISO Guide 34 certified reference material producer and by an A2LA accredited ISO 17025 calibration laboratory. Additional accreditations include ISO 31 (certificate content), and ISO 35 (statistical analysis), plus ISO 9001 DNV (certified quality manufacturer).

Accuracy at 25 °C is ±0.25 µmhos for 1, 5, and 10 µmhos solutions, and 0.25% for all other solutions; or the uncertainty shown on the certificate, whichever is greater. Each container is labeled for calibrating conductivity in micromhos (microsiemens), resistivity in megohms, and dissolved solids in ppm. Available in 100 ml (3.4 oz.) single-use polyethylene bottles or 473 ml (16 oz.) bottles.

Certifications: Meet test requirements for Federal, State, and local agencies, CAP, CLSI, ACS, CLIA and complies with AOAC 973.40, EPA 120.1, Standard Method 2510 (APHA, AWWA, WEF), ISO 7888, DIN 38404, ASTM D1125, USGS I-1780, USP 645, OIML R56, IUPAC, and for A2LA/NVLAP accreditations/ISO 9000 certifications.
Ordering information: Assortment 61161-356 includes one bottle each of the single-use standards, except the 5 µmhos solution (12777-832). All containers supplied with individual temperature compensation chart, calibration instructions, traceability information and Traceable® certificate.
Frequently Bought Together