VWR® Lab-Top Timer
# de catalogue 62344-910
Fournisseur: VWR International
Spécifications
- Cat. no.:62344-910
- W×H×D:8.3×8.3×1.9 cm (3¹/₄×3¹/₄׳/₄")
Spécifications
A propos de cet article
Designed for bench-use, this timer features any-angle viewing, large keys, and three timing modes.
- Traceable®
- For bench use
- Can display countdown, stopwatch, or time of day
Counts down from 24 hours to 1 second in one second increments. Countdown mode memory allows programming an 'instant replay' repeat time at the touch of a key. Also features a stopwatch and a time-of-day clock.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with a three-year AA alkaline battery.
Customers Who Bought This Also Bought