ContactUsLinkComponent
VWR* QC Timer*
VWR® QC Timer™ Timer
Catalog # 62344-902
Supplier:  VWR International
undefined
VWR® QC Timer™ Timer
Catalog # 62344-902
Supplier:  VWR International
Specifications
  • Cat. no.:
    62344-902
  • W×H×D:
    8.9×5.1×1.3 cm (3¹/₂×2×¹/₂")
Specifications
About this item
Ideal quality control countdown and alarm timer combines easy three-button operation with large 1.9 cm (³/₄") digits.
  • Traceable®
  • Ideal ISO 9000 timer
  • Countdown alarm
  • Magnet/clip on back
Can be set up to 19 hours, 59 minutes. Alarm sounds for one minute.
Certifications: A serial numbered certificate is provided from an ISO 17025 calibration laboratory accredited by A2LA to indicate instrument traceability to standards provided by the National Institute of Standards and Technology.
Ordering information: Supplied with three-year AAA alkaline battery.
Frequently Bought Together