VWR® Mini-Dolly Folding Cart
# de catalogue 61161-316
Fournisseur: VWR International
Spécifications
- Description:Mini-Dolly Folding Cart
- Cat. no.:61161-316
- Load Capacity:265 lbs.
Spécifications
A propos de cet article
Lightweight platform cart ideal for moving chemicals, carboys, computer paper, filing boxes, and scientific equipment.
- Easily folds for convenient storage
- Plastic guards protect walls
Eliminates lifting required by high-shelf carts and holds loads up to 120kg (265lbs.). The cart folds to a compact size for easy storage and features a nonskid 45x64.8 cm (173/4x251/2") platform. Plastic guards on all four sides protect walls, and elastic cords supplied with the cart are designed with quick hooks to help secure the load. Super-glide wheels swivel for complete maneuverability. Comfortable easy-grip handle extends to 87.6 cm (341/2"), ensuring a firm hold. The cart is constructed of high-impact ABS plastic and durable chrome-plated steel.
Customers Who Bought This Also Bought