VWR® Plate Roller
# de catalogue 60941-118
Fournisseur: VWR International
Spécifications
- Description:VWR® Plate Roller
- Cat. no.:60941-118
Spécifications
A propos de cet article
This 10.2 cm (4") soft rubber roller is designed for assuring tight and consistent application of adhesive films and foils to microplates.
- Tight and consistent application
- Secure seal around all microplate wells
Customers Who Bought This Also Bought