Order Entry
ContactUsLinkComponent
VWR* FILM, CLEAR ZONES, NONSTERILE CS50
VWR® Clear-Zone Pierceable Films for Robotics
Catalog # 60941-090
Supplier:  VWR International
VWR* FILM, CLEAR ZONES, NONSTERILE CS50
VWR® Clear-Zone Pierceable Films for Robotics
Catalog # 60941-090
Supplier:  VWR International

Specifications

  • Optical property:
    Transparent
  • Sterility:
    Non sterile
  • Description:
    Clear-Zone films
  • Thickness:
    96,6 µm
  • Cat. no.:
    60941-090

Specifications

About this item

With a polyethylene top layer and inert white polypropylene and acrylic adhesive sublayer, this easy-piercing sealing film allows direct sample recovery with single or multichannel pipettors and robotic probes.

  • Clear zone above each well is free of adhesive
  • Minimise sample exposure to adhesive
  • Dimensions: 14.3x7.9 cm (55/8x31/8")
  • Recommended temperature range −40 to +90 °C

The adhesive-free zone above each well prevents adhesive fouling of the pipette tip or robotic probe, minimising the amount of adhesive that comes into contact with the sample. Suitable for 96-well plates only.

Customers Who Bought This Also Bought