ContactUsLinkComponent
Identi-Plug® Plastic Foam Plugs, Jaece
Catalogue # 60882-205
Fournisseur:  Jaece
undefined
Identi-Plug® Plastic Foam Plugs, Jaece
Catalogue # 60882-205
Fournisseur:  Jaece
Supplier Number:  L800-E
Spécifications
  • Color:
    White
  • Description:
    White Foam Plugs
  • Cat. no.:
    60882-205
  • For Tube O.D.:
    46–65 mm
  • Supplier No.:
    L800-E
Spécifications
À propos de cet article
These reusable plastic white foam plugs are for use with test tubes and flasks.
  • Provides a snug fit, yet permits free air flow
  • Proven nontoxic to algae
  • Breathable and non-absorbent
  • Autoclavable
  • Chemical resistant
  • Springs back to original shape after repeated use
These plugs can also be used as a cleaner for lab pipettes, tubes, and vials, as well as industrial precision tubing.
Ordering information: Assorted Color Variety Pack includes 150 plugs each of the following assorted colors: red, green, yellow, charcoal, blue, and white.
Frequently Bought Together