Specifications
- Color:White
- Cat. no.:60828-760
Specifications
About this item
Safe-T-Flex caps feature a unique, flexible, over-locking design that locks securely on the outside of the tube lip and can be removed with one hand.
- Polyethylene construction
- Liquid-tight seal
- Color-coded for easy specimen identification
- Cap top can be labeled
- Self-locking caps hold firmly in any position
A tight seal ensures sample integrity and prevents spillage or evaporation of serum samples
Caps fit all varieties of blood collecting and disposable culture tubes. They are color-coded for easy specimen identification and to help guard against cross-contamination. Top of cap can also be numbered or labelled with indelible ink. Self-locking caps hold firmly in any position and keep contents secure while travelling. Liquid-tight seal withstands centrifugation, agitation, refrigeration, and freezing. Manufactured from polyethylene.
This special seal protects against the aerosols of highly infectious organisms such as TB and the HTLV-III virus.