PYREX® Disposable Culture Tubes, Rimless, Dispenser Pack and Bulk Pack, Corning
# de catalogue 60826-905
Fournisseur: Corning
Product available on GSA Advantage®
Spécifications
- Capacity:36 ml
- Ø:20 mm
- Description:Round-bottom, rimless culture tube
- Height:150 mm
- Supplier No.:99445-20
Spécifications
A propos de cet article
Made of borosilicate glass to reduce pH changes and contaminants potentially leached from soda-lime glass.
These one-use culture tubes are packaged for your convenience in easy-to-open, easy-to-use packs. The three small sizes come in a dispenser arrangement, and the larger sizes are bulk-packed.
Customers Who Bought This Also Bought