Black Phenolic Screw Caps for ACCUFORM® SSR™ Vials, Kimble Chase
Catalog # 60826-304
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap size:13-415
- Description:Phenolic Caps with PTFE-Faced/14B White Rubber Liner
- Cat. no.:60826-304
- Supplier No.:73802-13415