KIM-KAP® Caps, Disposable, Polypropylene, Kimble Chase
# de catalogue 60825-801
Fournisseur: DWK LIFE SCIENCES S. DE R.L. DE MX
Spécifications
- Color:Natural
- For Tube O.D.:18 mm
- Supplier No.:73660-18
Spécifications
A propos de cet article
These KIM caps are designed to fit culture tubes with outer diameters ranging from 13 mm to 38 mm.
- Autoclavable
- Color coded
Customers Who Bought This Also Bought