ContactUsLinkComponent
KIM-KAP* Polypropylene Cap
KIM-KAP® Caps, Disposable, Polypropylene, Kimble Chase
Catalog # 60825-787
KIM-KAP* Polypropylene Cap
KIM-KAP® Caps, Disposable, Polypropylene, Kimble Chase
Catalog # 60825-787
Specifications
  • Color:
    Natural
  • Cat. no.:
    60825-787
  • For Tube O.D.:
    13 mm
  • Supplier No.:
    73660-13
Specifications
About this item
These KIM caps are designed to fit culture tubes with outer diameters ranging from 13 mm to 38 mm.
  • Autoclavable
  • Color coded
Customers Who Bought This Also Bought