Order Entry
ContactUsLinkComponent
VWR* Culture Tube
VWR® Culture Tubes, Disposable, Borosilicate Glass
# de catalogue 60825-447
Fournisseur:  VWR International
VWR* Culture Tube
VWR® Culture Tubes, Disposable, Borosilicate Glass
# de catalogue 60825-447
Fournisseur:  VWR International

Spécifications

  • Capacity:
    34 ml
  • Ø:
    20 mm
  • Height:
    150 mm

Spécifications

A propos de cet article

These tubes are chemically clean and well-suited for general use in chemistry, blood banks, hematology, and other laboratory procedures.

Tubes are packed in lint-free packaging and are shrink-film sealed to prevent any contamination from outside sources.

  • Made of borosilicate glass 7.0
  • Fire-polished top
  • Well-rounded bottoms
  • Shrink-film sealed to prevent contamination

These tubes are chemically clean and well-suited for general use in chemistry, blood banks, hematology, and other laboratory procedures.

Each tube is annealed at temperatures over 426.7 °C (800 °F) to increase strength and reduce breakage.

Tubes are packed in lint-free packaging and are shrink-film sealed to prevent any contamination from outside sources.

Tubes feature smooth, well-rounded bottoms with no irregularities. Tops are fire-polished to eliminate rough edges.

Customers Who Bought This Also Bought