Syringes and Tubing for MICROLAB® 500 Series Dispensers/Diluters, Hamilton
# de catalogue 60375-555
Fournisseur: Hamilton
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Volume:5 ml
- Description:Model 1005 TLL Syringe
- Supplier No.:81520
Spécifications
A propos de cet article
For use with 25227-830 series dispensers/diluters.
Note: For dual dispenser models, use reagent (left side) syringes on both sides.
Customers Who Bought This Also Bought