ContactUsLinkComponent
900 Series MICROLITER* Removable Needle Syringe, Point Style 2
Microliter™ 900 Series Syringes, Hamilton
# de catalogue 60374-020
Fournisseur:  Hamilton
900 Series MICROLITER* Removable Needle Syringe, Point Style 2
Microliter™ 900 Series Syringes, Hamilton
# de catalogue 60374-020
Fournisseur:  Hamilton
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Volume:
    10 µl
  • Description:
    Model 901 Small RN Syringe
  • Supplier No.:
    80370
  • Barrel I.D.:
    0.019" (0.485 mm)
Spécifications
A propos de cet article
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
Each is manufactured to achieve the highest level of accuracy and precision possible. The 900 series syringe is the economical version of the 800 series. Attached to the flange of the syringe is a blue syringe holder that prevents heat transfer, dispense inaccuracies and plunger blow outs. Additionally it provides support to small volume plungers that are prone to bending during injection. The holder is permanently attached to the syringe barrel. This reduces the initial cost of the syringe but eliminates the possibility of replacement components. The 900 series syringes are ideal for dispensing volumes from 0.5 μL up to 10 μL.
Cemented Needle (N)- For low volume syringes, the needles are cemented into the glass syringe barrel at a point corresponding to the zero graduation mark. With this termination, dead volume is limited to the internal volume of the needle. Not autoclavable. Needle gauge is determined by the syringe volume and are not user-selectable.
Removable Needle (RN)- The needles are removable and are a Hamilton-specific design. The design allows the needles to seat precisely at the zero graduation mark of the syringe. Users can select the needle gauge, length, and point style to optimize the syringe for custom applications. Additionally, this termination allows for a removable needle without increasing the dead volume of the syringe and is ideal when there is a risk of the needle clogging. Autoclavable when disassembled. Repeated autoclaving will shorten syringe life.
Needle Point Style:
Point Style 2 - Sharp, beveled, curved, non-coring needle point recommended for septum penetration. Available gauges: 34-22.
Ordering information: If you are looking for a needle point or gauge that is available but not listed here, please call Avantor to place a special order by phone.
Customers Who Bought This Also Bought