ContactUsLinkComponent
Replacement Plunger Assembly N for Model 1705
Gastight® 1700 Series Syringe Replacement Plungers, Hamilton Company
# de catalogue 60373-638
Fournisseur:  Hamilton
Replacement Plunger Assembly N for Model 1705
Gastight® 1700 Series Syringe Replacement Plungers, Hamilton Company
# de catalogue 60373-638
Fournisseur:  Hamilton
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Hamilton Model 1705 Replacement Plunger
  • Cat. no.:
    60373-638
  • Supplier No.:
    1162-01
  • Termination:
    N, RN, TLL, C
Spécifications
A propos de cet article
Hamilton Company offers the most comprehensive selection of standard and custom syringes in the industry
These replacement plungers are for Hamilton 1700 series Gastight® syringes ranging in volume from 10 to 500 µl.
Each is manufactured to achieve the highest level of accuracy and precision possible.
Customers Who Bought This Also Bought