Accessories for Synthware Vacuum Adapters with Glass Stopcock
Catalog # 60005-162
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Replacement 2 mm glass plug
- Cat. no.:60005-162
- Supplier No.:S461230
Specifications
About this item
Kemtech all glass replacement straight bore stopcock.
- Borosilicate 3.3
- Straight bore
- 12/30 plug size
- 1:10 taper
To be used with our constant pressure retention kit.
Frequently Bought Together