Synthware Cannula, Stainless Steel, Kemtech America
Catalog # 60005-088
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Cat. no.:60005-088
- I.D.×O.D.:1.65×1.20 mm
- Gauge:16
- Needle Length:900 mm
- Supplier No.:C701636
Specifications
About this item
Designed for the transfer and handling of air sensitive materials.
- Stainless steel construction
- 16, 18, 20 and 22 guage available
- Various lengths available
Frequently Bought Together