ContactUsLinkComponent
SEPTUM PTFE/SILICONE 8MM
Synthware Septa, Teflon®-Lined Silicone Rubber
# de catalogue 60005-060
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
SEPTUM PTFE/SILICONE 8MM
Synthware Septa, Teflon®-Lined Silicone Rubber
# de catalogue 60005-060
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Ø:
    8 mm
  • For:
    Side port hickman still head
  • Material:
    Teflon®-lined silicone rubber
  • Supplier No.:
    R1408
Spécifications
A propos de cet article
Replacement PTFE lined silicone septums.
  • Silicone construction
  • Ptfe lined
For use with open top closures.
Customers Who Bought This Also Bought