Synthware Three Finger Double Adjustable Clamp, Kemtech America
# de catalogue 60005-034
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Small
- Grip Size:0 - 60 mm
- Supplier No.:W192203
Spécifications
A propos de cet article
Double adjustable clamp with PVC coated prongs.
- Adjustable
- Plastic coated fingers
- Round plastic tension screws
- Chrome plated
Clamps are constructed of chrome-plated Zinc-alloy. Small clamp accepts objects from 0 to 60 mm. Large clamp accepts object from 0 to 102 mm.
Customers Who Bought This Also Bought