[ST] Joint Clips
Catalog # 60004-952
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Plastic joint clip
- Cat. no.:60004-952
- ST Joint Size:29/42
- Supplier No.:CS002942
Specifications
About this item
Made of plastic.
- Heavy duty plastic construction
- Color coded
- Fit standard taper joints
Our heavy duty keck clips are perfect for all standard taper ground glass joints.
Customers Who Bought This Also Bought