[ST] Joint Clips
# de catalogue 60004-948
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Description:Plastic joint clip
- ST Joint Size:19/22
- Supplier No.:CS001922
Spécifications
A propos de cet article
Made of plastic.
- Heavy duty plastic construction
- Color coded
- Fit standard taper joints
Our heavy duty keck clips are perfect for all standard taper ground glass joints.
Customers Who Bought This Also Bought