Synthware PTFE Flask Length Stopper, Kemtech America
Catalog # 60004-882
Supplier: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
- Description:Synthware PTFE flask length stopper
- Cat. no.:60004-882
- Joint Size:#38
- Supplier No.:S120038
Specifications
About this item
Stoppers features PTFE (Polytetrafluoroethylene) handle for easy griping.
- Teflon® construction
- Standard joint sizes available
Ideal for use in flasks.
Customers Who Bought This Also Bought