ContactUsLinkComponent
STOPPER P-H SOLID 24/40
Synthware Solid Pennyhead [ST] Stoppers, Glass, Kemtech America
# de catalogue 60004-824
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
STOPPER P-H SOLID 24/40
Synthware Solid Pennyhead [ST] Stoppers, Glass, Kemtech America
# de catalogue 60004-824
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • ST Joint No.:
    24/40
  • Supplier No.:
    S292440
Spécifications
A propos de cet article
Solid glass penny head stopper.
  • Heavy wall Borosilicate construction
  • Male ground glass joint
  • Penny head design
  • Solid
Penny head design allows for easy removal of the stopper.
Customers Who Bought This Also Bought