Synthware Solid Pennyhead [ST] Stoppers, Glass, Kemtech America
# de catalogue 60004-820
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- ST Joint No.:14/20
- Supplier No.:S291420
Spécifications
A propos de cet article
Solid glass penny head stopper.
- Heavy wall Borosilicate construction
- Male ground glass joint
- Penny head design
- Solid
Penny head design allows for easy removal of the stopper.
Customers Who Bought This Also Bought