Synthware Syringe Needles, Stainless Steel, Kemtech America
# de catalogue 60004-796
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Length:304.8 mm (12.00")
- O.D.:1.6 mm
- Supplier No.:S380230
Spécifications
A propos de cet article
Stainless steel needles are designed with extra length for solvent retrieval.
- Stainless steel construction
- Luer lock design
These long needles are ideal for solvent retrieval.
Customers Who Bought This Also Bought