Order Entry
ContactUsLinkComponent
VACUUM TRAP 500ML
Synthware Solvent Removal Vacuum Trap, Kemtech America
# de catalogue 60004-714
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
VACUUM TRAP 500ML
Synthware Solvent Removal Vacuum Trap, Kemtech America
# de catalogue 60004-714
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise

Spécifications

  • Capacity:
    500 ml
  • Description:
    Vacuum trap, 130×310 mm
  • Supplier No.:
    V120500

Spécifications

A propos de cet article

Trap is designed for large volume solvent removal by vacuum.

  • 45/50 joint connects two components
  • Heavy wall borosilicate construction

With [ST] 40/35 joint. Center tube connection has an ext. Ø of 12 mm. The side tube connection has an ext. Ø of 10 mm.

Ordering information: Trap 60004-714 fits a 1900 ml Dewar flask. Trap 60004-716 fits a 4300 ml Dewar flask.
Customers Who Bought This Also Bought