Synthware Flask, Filtering, Microscale, Kemtech America
# de catalogue 60004-226
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Capacity:50 ml
- Supplier No.:F150050
Spécifications
A propos de cet article
Filtering flask for microscale applications.
- Microscale
- Heavy wall Borosilicate construction
- Beaded top rim
- Barbed side arm
Top is tooled for a #2 neoprene filter adapter or #3 rubber stopper.
Customers Who Bought This Also Bought