Synthware Conical Reaction Vials, Heavy Wall
# de catalogue 60004-170
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Cap included:Yes
- Capacity:5.0 ml
- Description:14/10 joint
- Supplier No.:V131005
Spécifications
A propos de cet article
Graduated.[ST] Joint No.: 14/10.
- Heavy wall Borosilicate construction
- Threaded Ground glass joint
- Graduated
- Cap included
This heavy wall reaction vial is great for microscale reactions.
Ordering information: For replacement caps, Viton® O-rings, or septa, contact your Avantor representative.
Delivery: Supplied with one compression cap, one Viton® O-ring, and one fluoropolymer resin-faced silicone rubber septum.
Customers Who Bought This Also Bought