ContactUsLinkComponent
VIAL CONICAL RXN 14/10 5ML
Synthware Conical Reaction Vials, Heavy Wall
# de catalogue 60004-170
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
VIAL CONICAL RXN 14/10 5ML
Synthware Conical Reaction Vials, Heavy Wall
# de catalogue 60004-170
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Cap included:
    Yes
  • Capacity:
    5.0 ml
  • Description:
    14/10 joint
  • Supplier No.:
    V131005
Spécifications
A propos de cet article
Graduated.[ST] Joint No.: 14/10.
  • Heavy wall Borosilicate construction
  • Threaded Ground glass joint
  • Graduated
  • Cap included
This heavy wall reaction vial is great for microscale reactions.
Ordering information: For replacement caps, Viton® O-rings, or septa, contact your Avantor representative.
Delivery: Supplied with one compression cap, one Viton® O-ring, and one fluoropolymer resin-faced silicone rubber septum.
Customers Who Bought This Also Bought