ContactUsLinkComponent
FLASK RB 1N 14/20 100ML
Synthware Round-Bottom Flasks, Single-Neck, Kemtech America
# de catalogue 60002-298
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
FLASK RB 1N 14/20 100ML
Synthware Round-Bottom Flasks, Single-Neck, Kemtech America
# de catalogue 60002-298
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Capacity:
    100 ml
  • ST Joint No.:
    14/20
  • Cat. no.:
    60002-298
  • Supplier No.:
    F301100
Spécifications
A propos de cet article
Heavy wall design.
  • Heavy wall borosilicate construction
  • Ground glass joint
  • Various sizes available
Hand-blown from tubing to ensure uniform wall thickness.
Customers Who Bought This Also Bought