Synthware Claisen Adapter, Kemtech America
# de catalogue 60000-658
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- ST Joint:14/20
- Cat. no.:60000-658
- Supplier No.:A371420
Spécifications
A propos de cet article
This Claisen-style adapter provides dual entry into a variety of flasks.
- Allows dual entry
- Use with a variety of flasks
The upper joints are outer [ST] and the bottom joint is an inner [ST].
Customers Who Bought This Also Bought