Synthware Distilling Trap Adapters, Kemtech America
# de catalogue 60000-406
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Capacity:100 ml
- Cat. no.:60000-406
- Bottom [ST] Joint Size:24/40
- Top ST Joint Size:[ST] 24/40
- Supplier No.:A264100
Spécifications
A propos de cet article
Distilling trap adapter is placed above flask to prevent carryover of reactions that have a tendency to foam.
- Top [ST] joint
- Bottom [ST] joint
Adapter features an upper [ST] joint size of 24/40 and capacity of 100 ml.
Customers Who Bought This Also Bought