ContactUsLinkComponent
TRAP DIST 24/40-24/40 100ML
Synthware Distilling Trap Adapters, Kemtech America
# de catalogue 60000-406
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
TRAP DIST 24/40-24/40 100ML
Synthware Distilling Trap Adapters, Kemtech America
# de catalogue 60000-406
Fournisseur:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
  • Capacity:
    100 ml
  • Cat. no.:
    60000-406
  • Bottom [ST] Joint Size:
    24/40
  • Top ST Joint Size:
    [ST] 24/40
  • Supplier No.:
    A264100
Spécifications
A propos de cet article
Distilling trap adapter is placed above flask to prevent carryover of reactions that have a tendency to foam.
  • Top [ST] joint
  • Bottom [ST] joint
Adapter features an upper [ST] joint size of 24/40 and capacity of 100 ml.
Customers Who Bought This Also Bought