Synthware Reducing Adapters, Kemtech America
# de catalogue 60000-048
Fournisseur: Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Spécifications
- Cat. no.:60000-048
- Bottom [ST] Joint Size:45/50
- Top ST Joint Size:[ST] 24/40
- Supplier No.:A202445
Spécifications
A propos de cet article
Reducing adapters are used for connecting dissimilar [ST] joints.
- Top outer [ST] joint
- Bottom inner [ST] joint
Features an outer [ST] joint at the top and inner [ST] joint at the bottom.
Customers Who Bought This Also Bought