Order Entry
Canada
ContactUsLinkComponent
Stomacher® Bags for Stomacher® 400 Lab Blender Series, Seward
Catalogue # 58976-930
undefined
Stomacher® Bags for Stomacher® 400 Lab Blender Series, Seward
Catalogue # 58976-930
Supplier Number:  030020251

Spécifications

  • Description:
    Filter bags for Stomacher® 400
  • Cat. no.:
    58976-930
  • Supplier No.:
    030020251

Spécifications

À propos de cet article

The Stomacher® Bag range has been designed to fulfil all applications from sample blending, straining and storage, including bags designed for extensive blending (processes requiring more than 15 minutes on the machine) and small samples for life science applications.

  • Made from high quality food grade virgin LDPE granule extruded to 65 µm film thickness
  • Withstand extensive blending without tears or perforations
  • Low cost option for sample preparation with no risk of cross contamination between samples
  • Irradiated sterile

Bags for the Stomacher® 400 have a capacity of 80 to 400 ml. Compatible with all lab blenders and homogenizers.

Ordering information: Irradiation certificates are available on request.
Frequently Bought Together