Stomacher® Bags for Stomacher® 400 Lab Blender Series, Seward
# de catalogue 58976-921
Fournisseur: Seward Laboratory System
Spécifications
- Description:Standard bags for Stomacher® 400
- Cat. no.:58976-921
- Supplier No.:BA6041
Spécifications
A propos de cet article
The Stomacher® Bag range has been designed to fulfil all applications from sample blending, straining and storage, including bags designed for extensive blending (processes requiring more than 15 minutes on the machine) and small samples for life science applications.
- Made from high quality food grade virgin LDPE granule extruded to 65 µm film thickness
- Withstand extensive blending without tears or perforations
- Low cost option for sample preparation with no risk of cross contamination between samples
- Irradiated sterile
Bags for the Stomacher® 400 have a capacity of 80 to 400 ml. Compatible with all lab blenders and homogenizers.
Ordering information: Irradiation certificates are available on request.
Customers Who Bought This Also Bought