To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
- Dimensions:61W x 61L cm (24 x 24")
- Description:Steri-Wrap® I Single Use Wrap
- Cat. no.:58752-920
- Supplier No.:311024
Spécifications
A propos de cet article
Steri-Wrap® I is a medium weight wrap, ideal for general hospital use in steam and ethylene oxide sterilization cycles.
- Cellulose based wrap for Steam and EO sterilization
- Efficient microbial barrier, high-level of protection against fluids and particles
- Highly permeable meaning no wet packs
- Variety of strength of fabric to meet your needs
Steri-Wrap® I is cellulose based sterilization wrap available in three grades for keeping packages safe and secure during the sterilization process. Steri-Wrap® I is a medium weight wrap, ideal for general hospital use. Steri-Wrap® I is designed to be virtually lint free with tightly anchored fibers. Wrap is reversible and features excellent liquid repellency.