Accessories for VWR® 8" Sieves
# de catalogue 57334-652
Fournisseur: VWR International
Spécifications
- Description:Full Height Sieve Pan, Stainless Steel
- Cat. no.:57334-652
Spécifications
A propos de cet article
VWR® 8" diameter (203.2 mm ) durable pans and covers fit all VWR® 8" diameter sieves.
- Highly durable construction
- Brass or stainless steel fabrication
VWR® 8" diameter (203.2 mm ) durable pans and covers fit all VWR® 8" diameter sieves.
Half height (1") or full height (2") pans are available in brass or stainless steel. Pans with extended rims can be used for running multiple tests simultaneously. Sieve brushes are available as accessories.
Customers Who Bought This Also Bought