Absorbent Laboratory Countertop Paper, TIDI®
# de catalogue 54110-528
Fournisseur: Tidi Products
Small Business Enterprise
Spécifications
- Colour:White
- Description:Flat Sheet
- Size:30.5W x 50.8L cm (12” x 20”)
- Cat. no.:54110-528
- Supplier No.:980982
Spécifications
A propos de cet article
This paper is made of thick, absorbent double re-crepe (DRC) material for a soft, cloth-like feel.
- Pebble embossed for added cushioning
- Poly-coated backing provides fluid holdout
Customers Who Bought This Also Bought