SP Bel-Art Poxygrid® Microliter Pipettor Racks, Bel-Art Products, a part of SP
# de catalogue 53577-145
Fournisseur: Bel-Art Products, a Part of SP
Spécifications
- Dimensions:22.9W x 12.7D x 17.1H cm (9 x 5 x 63/4")
- Description:For Finnpipette®, Eppendorf®, and Other Popular Brand Single-Channel Pipettors
- Cat. no.:53577-145
- Supplier No.:189620000
Spécifications
A propos de cet article
Store pipettors while protecting pipettor tips in these epoxy-coated aluminum racks that provide quick, easy access for more efficient work.
- Epoxy-coated aluminum rack construction
- Hold a range of pipettors
- Suitable for wall mounting or free standing
Able to hold up to 10 round-shaped pipettors such as Finnpipette®, Socorex®, Eppendorf® and other popular brands in two stepped rows of 5 each. This rack also has end holes for the Pipet-Aid® pipette filler. A second lower support row has smaller holes that accept the tapered ends of the pipettors to keep them vertical.
Customers Who Bought This Also Bought