Finntip™ Pipette Tips, Thermo Scientific
Catalog # 53515-982
Supplier: Thermo Fisher Scientific
Specifications
- Volume:500 - 5000 µl
- Packed:5×54/Rack
- Cat. no.:53515-982
- Sterile:+
- Tip Style:Finntip™ 5 mL
- Supplier No.:9402073
Specifications
About this item
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
- Made of high-quality polypropylene
- Autoclavable
- Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought