ContactUsLinkComponent
PIPETTE TIP FINNTP S 5ML CS270
Finntip™ Pipette Tips, Thermo Scientific
Catalog # 53515-982
Finntip™ Pipette Tips, Thermo Scientific
Catalog # 53515-982
Specifications
  • Volume:
    500 - 5000 µl
  • Packed:
    5×54/Rack
  • Cat. no.:
    53515-982
  • Sterile:
    +
  • Tip Style:
    Finntip™ 5 mL
  • Supplier No.:
    9402073
Specifications
About this item
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
  • Made of high-quality polypropylene
  • Autoclavable
  • Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought