ContactUsLinkComponent
Finntip* Pipet Tips
Finntip™ Pipette Tips, Thermo Scientific
Catalog # 53515-954
Finntip™ Pipette Tips, Thermo Scientific
Catalog # 53515-954
Specifications
  • Volume:
    0.2 - 10 µl
  • Packed:
    1000/Bag
  • Cat. no.:
    53515-954
  • Tip Style:
    Finntip™ 10
  • Supplier No.:
    9400310
Specifications
About this item
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
  • Made of high-quality polypropylene
  • Autoclavable
  • Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought