Finntip™ Pipette Tips, Thermo Scientific
Catalog # 53515-954
Supplier: Thermo Fisher Scientific
Specifications
- Volume:0.2 - 10 µl
- Packed:1000/Bag
- Cat. no.:53515-954
- Tip Style:Finntip™ 10
- Supplier No.:9400310
Specifications
About this item
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
- Made of high-quality polypropylene
- Autoclavable
- Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought