ContactUsLinkComponent
Finntip Stepper, 25mL, Nonsterile
Finntip Stepper Tips, Thermo Scientific
# de catalogue 53515-932
Finntip Stepper Tips, Thermo Scientific
# de catalogue 53515-932
Spécifications
  • Volume:
    500 µl, 1000 µl, 1500 µl, 2000 µl, 2500 µl
  • Tip Style:
    Finntip Stepper 25 mL
  • Supplier No.:
    9404220
Spécifications
A propos de cet article
Finntip stepper tips are the perfect fit with Thermo Scientific Finnpipette stepper pipetters.
The finntip stepper tips are plunger-style tips for positive displacement pipetting. Seven sizes are available in sterile and non sterile versions.
Customers Who Bought This Also Bought