ContactUsLinkComponent
Finntip Stepper, 2.50mL, Sterile
Finntip Stepper Tips, Thermo Scientific
# de catalogue 53515-924
Finntip Stepper, 2.50mL, Sterile
Finntip Stepper Tips, Thermo Scientific
# de catalogue 53515-924
Spécifications
  • Volume:
    50 µl, 100 µl, 150 µl, 200 µl, 250 µl
  • Sterile:
    +
  • Tip Style:
    Finntip Stepper 2.5 mL
  • Supplier No.:
    9404193
Spécifications
A propos de cet article
Finntip stepper tips are the perfect fit with Thermo Scientific Finnpipette stepper pipetters.
The finntip stepper tips are plunger-style tips for positive displacement pipetting. Seven sizes are available in sterile and non sterile versions.
Customers Who Bought This Also Bought