Finntip™ Pipette Tips, Thermo Scientific
# de catalogue 53515-888
Fournisseur: MOLECULAR BIOPRODUCTS INC MX
Spécifications
- Volume:1000 - 5000 µL
- Packed:500/Bag
- Tip Style:Finntip™ 5 mL
- Supplier No.:9402030
Spécifications
A propos de cet article
These high-quality pipette tips are specifically designed to fit Finnpipette® pipettors, but are also compatible with most pipettor brands.
- Made of high-quality polypropylene
- Autoclavable
- Chemical-resistant
Tips in refill trays can be used to quickly refill existing racks.
Customers Who Bought This Also Bought