Wiretrol® II Disposable Micropipettes, Drummond
# de catalogue 53507-432
Fournisseur: Drummond
Spécifications
- Capacity:25/50 µl
- Color:Double White
- Description:Wiretrol® II Micropipet
- Cat. no.:53507-432
- Supplier No.:5-000-2050
Spécifications
A propos de cet article
This economical positive-displacement micropipette features fingertip control.
- Dual graduations for non-contact delivery
- Ideal for a wide variety of applications including both high- and low-viscosity liquids and corrosives
- Economical
The Wiretrol II provides the accuracy and convenience of the Wiretrol I series with dual graduations and a shorter plunger for non-contact delivery.
Customers Who Bought This Also Bought