VWR® Pipette Filler, Synthetic Rubber
Catalog # 53502-205
Supplier: VWR International
Product available on GSA Advantage®
Specifications
- Color:Red
- Cat. no.:53502-205
Specifications
About this item
Pipette fillers do away with the frequently hazardous practice of drawing liquids into pipettes by mouth.
- No metal parts to corrode
For use with pipettes from 2 to 50 ml. Made of synthetic rubber with glass ball valves.
Ordering information: Supplied with an adapter for capacities greater than 20 ml.
Customers Who Bought This Also Bought