VWR® Repeating Dispensers
# de catalogue 53481-406
Fournisseur: VWR International
Spécifications
- Volume:15 ml
Spécifications
A propos de cet article
Dispensers are recommended for multiple test procedures as well as dispensing strong acids, alkalies, and volatile reagents.
- Accurate to ±2%
- Capable of rapid repeat transfers of fixed volumes
- Easy filling and dispensation
Dispensers are recommended for multiple test procedures as well as dispensing strong acids, alkalies, and volatile reagents.
The calibrated pipet heads automatically fill and deliver contents by simple pouring action for rapid repeat transfers of fixed volumes of solutions. To fill, the pipet head is tilted backwards. Excess liquid returns to the flask automatically. Tilting dispenser forward delivers the calibrated volume. Pipet heads ordered separately have [ST] 24/40 inner joint.
Ordering information: A complete unit consists of an interchangeable glass pipet head, Erlenmeyer flask with [ST] 24/40 ground joint, and poly joint clamp.
Customers Who Bought This Also Bought