ContactUsLinkComponent
Wiretol II Capillary Microdispenser
Wiretrol® II Disposable Micropipettes, Drummond
# de catalogue 53480-655
Fournisseur:  Drummond
Wiretrol® II Disposable Micropipettes, Drummond
# de catalogue 53480-655
Fournisseur:  Drummond
Spécifications
  • Capacity:
    100/200 µl
  • Color:
    Blue
  • Description:
    Wiretrol® II Micropipet
  • Cat. no.:
    53480-655
  • Supplier No.:
    5-000-2200
Spécifications
A propos de cet article
This economical positive-displacement micropipette features fingertip control.
  • Dual graduations for non-contact delivery
  • Ideal for a wide variety of applications including both high- and low-viscosity liquids and corrosives
  • Economical
The Wiretrol II provides the accuracy and convenience of the Wiretrol I series with dual graduations and a shorter plunger for non-contact delivery.
Customers Who Bought This Also Bought