Order Entry
Canada
ContactUsLinkComponent
PYREX* Volumetric Pipets, Color-Coded, Class A, Tempered Top and Tip
PYREX® Volumetric Pipettes, Color-Coded, Class A, Corning
Catalog # 53044-070
Supplier:  Corning
Product available on GSA Advantage®
   Low Manufacturing Impact
undefined
PYREX® Volumetric Pipettes, Color-Coded, Class A, Corning
Catalog # 53044-070
Supplier:  Corning
Supplier Number:  7100-3
Product available on GSA Advantage®
   Low Manufacturing Impact

Specifications

  • Capacity:
    3 ml
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Environmental certification:
    I-RECs, ISO 14001 - Monterrey
  • Cat. no.:
    53044-070
  • Tolerance:
    ±0.01 ml
  • Band Color:
    Black
  • Supplier No.:
    7100-3

Specifications

About this item

Colored markings remain sharply legible for the life of pipet.

  • Reusable, rugged construction
  • Color-coded
  • Colored enamel graduations and markings

Ruggedly constructed and properly proportioned, pipettes have accuracy tolerances to meet or exceed the strict requirements for Class A Volumetric Ware as indicated by ASTM E969.

Frequently Bought Together