VWR® Permanent Marking Pens
# de catalogue 52877-886
Fournisseur: VWR International
Spécifications
- Color:Blue
- Cat. no.:52877-886
Spécifications
A propos de cet article
Perfect marking pens for small lab samples, slides, beakers, sample bags, and more.
- Available in a wide variety of colors
- Permanent ink is water- and fade-proof
- Airtight ring seal cap
Four brilliant colors produce precise lines of 0.3mm width
Write on glass, film, plastic, metal, porcelain, wood, rubber, and paper.
Permanent ink dries in a split second and is water- and fade-proof. Airtight ring seal cap prevents dry-out.
Ordering information: Assortment pack consists of six black, two red, two blue, and two green pens.
Customers Who Bought This Also Bought